Learn More
Phosphorus lump, red, Puratronic™, 99.999+% (metals basis)
Product Code. 11376497
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376497 1 g 1g
11356497 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11376497 Supplier Thermo Scientific Alfa Aesar Supplier No. 000248.03
 Restricted Item
Verfügbar ab: 21-12-2026
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
CAS: 7723-14-0
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7723-14-0
Physical Form Solid
Quantity 1 g
Percent Purity ≥99.999%
compliance-icons
Product Identifier
  • Phosphorus lump, red
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H228-Flammable solid.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..