Learn More
Phosphorus powder, -100 mesh, red amorphous, 98% (metals basis)
Product Code. 11381358
Change view
Click to view available options
Quantity:
500 g
4 x 500 g
Unit Size:
2000g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381358 4 x 500 g 2000g
11361358 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11381358 Supplier Thermo Scientific Alfa Aesar Supplier No. 010281.57
 Restricted Item
Verfügbar ab: 21-12-2026
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
CAS: 7723-14-0
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7723-14-0
Physical Form Solid
Quantity 4 x 500 g
Percent Purity 98.9%