missing translation for 'onlineSavingsMsg'
Learn More

PI 103 hydrochloride, Tocris Bioscience™

Inhibitor of PI 3-kinase, mTOR and DNA-PK

Marke:  Tocris (Bio-Techne) 293010

Weitere Details : Netto-Gewicht : 0.00001kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412074

  • 271.93 € / 10 Milligramm


Voraussichtlicher Liefertermin 26-03-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Inhibitor of DNA-PK, PI 3-kinase (p110α) and mTOR (IC 50 values are 2, 8, 20, 26, 48, 83, 88, 150, 850, 920, ∽ 1000 and 2300 nM for DNA-PK, p110α, mTORC1, PI 3-KC2β, p110δ, mTORC2, p110β, p110γ, ATR, ATM, PI 3-KC2α and hsVPS34 respectively). Inhibits growth of human tumor xenografts in mice in vivo. Induces autophagosome formation in glioma cells.
Spezifikation

Chemische Identifikatoren

371935-79-4
384.82
pi 103 hydrochloride, pi-103 hydrochloride, pi-103 hydrochloride, pi103 hydrochloride, pubchem22442, pi-103, hydrochloride, c19h16n4o3.clh, pi 103 hydrochloride;pi103 hydrochloride, 3-4-morpholinopyrido 3',2':4,5 furo 3,2-d pyrimidin-2-yl phenol hydrochloride, 3-4-4-morpholinylpyrido 3',2':4,5 f uro 3,2-d pyrimidin-2-yl phenol hydrochloride
3-(4-morpholin-4-ylpyrido[2,3]furo[2,4-b]pyrimidin-2-yl)phenol;hydrochloride
C19H17ClN4O3
XSQMYBFFYPTMFE-UHFFFAOYSA-N
16739368
C1COCCN1C2=NC(=NC3=C2OC4=C3C=CC=N4)C5=CC(=CC=C5)O.Cl

Spezifikation

PI 103 hydrochloride
371935-79-4
pi 103 hydrochloride, pi-103 hydrochloride, pi-103 hydrochloride, pi103 hydrochloride, pubchem22442, pi-103, hydrochloride, c19h16n4o3.clh, pi 103 hydrochloride;pi103 hydrochloride, 3-4-morpholinopyrido 3',2':4,5 furo 3,2-d pyrimidin-2-yl phenol hydrochloride, 3-4-4-morpholinylpyrido 3',2':4,5 f uro 3,2-d pyrimidin-2-yl phenol hydrochloride
C1COCCN1C2=NC(=NC3=C2OC4=C3C=CC=N4)C5=CC(=CC=C5)O.Cl
384.82
384.82
10mg
C19H17ClN4O3
XSQMYBFFYPTMFE-UHFFFAOYSA-N
3-(4-morpholin-4-ylpyrido[2,3]furo[2,4-b]pyrimidin-2-yl)phenol;hydrochloride
16739368
>98%
Videos