Learn More
Thermo Scientific™ Pierce™ Rinderserumalbumin Standards
High-quality, calibrated BSA solutions (2mg/mL) to serially dilute to create protein assay standard curves to accurately measure protein concentration.
81.75 € - 143.00 €
Produkte 3
Produktcode Marke Bezeichnung Menge. Preis Menge & Verfügbarkeit  
Produktcode Marke Bezeichnung Menge. Preis Menge & Verfügbarkeit  
10531674 Thermo Scientific™
23208
Pierce Standard-Set vorverdünntes Rinderserumalbumin 7 x 3.5 ml 143.00 €
24.50 Milliliter Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Auf Lager: 15
Zum Warenkorb hinzufügen
10443834 Thermo Scientific™
23210
Pierce 2 mg/ml Rinderserumalbumin Standard 50 ml 82.50 €
50 Milliliter Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Auf Lager: 21
Zum Warenkorb hinzufügen
11811345 Thermo Scientific™
23209
Pierce Rinderserumalbumin Standard Ampullen, 2 mg/ml 10 x 1 ml 81.75 €
10 Stück Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Auf Lager: 352
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
These bovine serum albumin (BSA) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BSA is the universally accepted reference protein for total protein quantitation. The albumin standard is precisely formulated at 2mg/mL in an ultrapure 0.9% sodium chloride (saline) solution. The concentration of the stock solution is calibrated by direct comparison to purified BSA (Fraction V) from the National Institute of Standards and Technology (NIST™). The product is available in three convenient package formats: 1mL glass ampules, 50mL polypropylene bottles, and complete sets containing seven ready-to-use dilutions.
Highlights:
  • Universal – recognized as the industry standard for protein quantitation in colorimetric protein assays
  • Pure and stable – supplied in ultrapure 0.9% saline solution with 0.05% sodium azide; room temperature stable
  • Accurate and consistent – precisely formulated at 2.00 +/-0.03mg/mL compared to a NIST reference
  • Convenient – choose 1mL ampules, a 50mL bottle, or a complete set containing seven ready-to-use dilutions
Includes:
2mg/mL BSA solution (Part No. 23209, 23210) or 7-solution set of different concentrations (Part No. 23208)
Recommended for:
Protein assay quantitation standard; Protein recovery control for desalting and other column procedures; General calibration of spectrophotometer UV-lamp (A280)