Learn More
Thermo Scientific™ Pierce™ Lane Marker Probenpuffer
Ready-to-use 5X SDS-PAGE sample loading buffers available in reducing and nonreducing formulations, both with a pink dye buffer-front marker.
74.50 € - 81.50 €
Spezifikation
Menge. 5 ml
Ausreichend für 500 bis 2500 Proben (25 ml insgesamt) für SDS-PAGE
Mehr Spezifikationen ansehen
Produkte 2
Produktcode Marke Bezeichnung Preis Menge & Verfügbarkeit  
Produktcode Marke Bezeichnung Preis Menge & Verfügbarkeit  
11809340 Thermo Scientific™
39000
Pierce Lane Marker Reducing Sample Puffer (5X) 81.50 €
5 Milliliter
Auf Lager: 171
Zum Warenkorb hinzufügen
Alternative Produkte anzeigen »
11819340 Thermo Scientific™
39001
Pierce Lane Marker Non-Reducing Sample Puffer (5X) 74.50 €
5 Milliliter
Auf Lager: 20
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Lane Marker Reducing and Non-Reducing Sample Buffers are convenient and ready to use for SDS-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blotting. Formulated as 5X stocks rather than the traditional 2X stocks, they enable a larger volume of protein solution to be included in the sample that is loaded in each well. Particularly unique in these buffers is the use of a bright pink tracking dye instead of the traditional bromophenol blue dye. This pink dye marks not only the dye front in the gel but also transfers and permanently binds to nitrocellulose membranes. Consequently, the dye can be used to assess transfer efficiency and align cut membranes. The Reducing Sample Buffer contains DTT as the reducing agent, eliminating the repugnant odor associated with mercaptoethanol-containing buffers.
Lane Marker Sample Buffers are easy to use. Simply mix one part Sample Buffer with four parts protein sample, heat denature, and then load the gel sample wells. Visualize progress of the electrophoresis run by monitoring the location of the pink dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques, as well as Western blotting procedures.
Highlights:
  • Easy to see – bright pink dye marks the buffer front, clearly indicating the progress of the electrophoresis run
  • Concentrated – 5X stocks enable a greater range of protein sample volumes to be used for electrophoresis
  • Transferable marker dye – pink dye transfers with protein to nitrocellulose membrane, enabling verification of transfer direction and orientation of the blot
  • Flexibility in SDS-PAGE applications – available in two ready-to-use formulations for reducing or non-reducing denaturing SDS polyacrylamide electrophoresis
Spezifikation
Spezifikation
5 ml
500 bis 2500 Proben (25 ml insgesamt) für SDS-PAGE