Learn More
Pifithrin-μ, Tocris Bioscience™
Inhibitor of p53-mitochondrial binding
Marke:  Tocris (Bio-Techne) 265350
Weitere Details : Netto-Gewicht : 0.01000kg
Menge. 50mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411664
  • 469.32 € / 50 Milligramm
Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Inhibits p53 binding to mitochondria by reducing its affinity for antiapoptotic proteins Bcl-2 and Bcl-XL. Displays no effect on the transactivational or cell cycle checkpoint control function of p53. Potentially increases reprogramming efficiency of human somatic cells to induced pluripotent stem cells (iPSCs) by silencing p53. Reduces cell death induced by γ-radiation in vitro and protects mice from doses of radiation that cause lethal hematopoietic syndrome. Selectively inhibits heat shock protein 70 (HSP70) activity.
Spezifikation
Chemische Identifikatoren
64984-31-2
181.209
pifithrin-mu, pifithrin-u, pifithrin-, i, 2-phenylacetylenesulfonamide, phenylethynesulfonamide, phenylacetylenylsulfonamide, 2-phenylethyne-1-sulfonamide, pifithrin mu, pftmu, pifithrin-micro
2-phenylethynesulfonamide
C8H7NO2S
ZZUZYEMRHCMVTB-UHFFFAOYSA-N
327653
C1=CC=C(C=C1)C#CS(=O)(=O)N
Spezifikation
Pifithrin-μ
64984-31-2
pifithrin-mu, pifithrin-u, pifithrin-, i, 2-phenylacetylenesulfonamide, phenylethynesulfonamide, phenylacetylenylsulfonamide, 2-phenylethyne-1-sulfonamide, pifithrin mu, pftmu, pifithrin-micro
C1=CC=C(C=C1)C#CS(=O)(=O)N
181.209
181.21
50mg
C8H7NO2S
ZZUZYEMRHCMVTB-UHFFFAOYSA-N
2-phenylethynesulfonamide
327653
>99%
Videos