Learn More
PIGB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558993
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QRGTLDVMSH IQKVCYNNPN KSSASIFIMM PCHSTPYYSH VHCPLPMRFL QCPPDLTGKS HYLDEAD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
PIGB Polyclonal antibody specifically detects PIGB in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
PIGB | |
Polyclonal | |
Unconjugated | |
PIGB | |
dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 3; GPI mannosyltransferase III; GPI-MT-III; involved in GPI-anchor biosynthesis; phosphatidylinositol glycan anchor biosynthesis class B; phosphatidylinositol glycan anchor biosynthesis, class B; phosphatidylinositol glycan, class B; phosphatidylinositol-glycan biosynthesis class B protein; Pigb; Pig-b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
315807, 55981, 9488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92521, Q9JJQ0 | |
PIGB | |
Recombinant protein corresponding to Human PIGB | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only