missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pigmentrot 146 (Technische Qualität), TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Pigment Red 146 (Technical Grade) |
| CAS | 5280-68-2 |
| Summenformel | C33H27ClN4O6 |
| Menge | 1 g |
| Synonym | Renol Carmine FBB-H, Sandye Super Pink FBL, Sanyo Pigment Pink 4602, Seikafast Carmine 3870, Sunbrite Red 146, Symuler Fast Red 4195, Symuler Fast Red 4580, Symuler Fast Red 4590, Vulcan Fast Carmine FBB, N-(4-Chloro-2,5-dimethoxyphenyl)-3-hydroxy-4-[2-[2-methoxy-5-[(phenylamino)carbonyl]phenyl]diazenyl]-2-naphthalenecarboxamide, C.I. 12485, C.I. PR 146, C.I. Pigment Red 146, Fuji Fast Carmine 550, Helio Fast Carmine BB, Hydrocolor Red, Karmine FBB 02, Lionol Red 5620, Microlith Red 3R-A, Naphtol Carmine 4635C, PR 146, Permanent Carmine FBB, Permanent Carmine FBB 02, Permanent Carmine FBB 02JP, Permanent Carmine FBB Extra, Pigment Pink 4602 |
| InChI-Formel | InChI=1S/C33H27ClN4O6/c1-42-27-14-13-20(32(40)35-21-10-5-4-6-11-21)16-26(27)37-38-30-22-12-8-7-9-19(22)15-23(31(30)39)33(41)36-25-18-28(43-2)24(34)17-29(25)44-3/h4-18,39H,1-3H3,(H,35,40)(H,36,41) |
| SMILES | COc1cc(NC(=O)c2cc3ccccc3c(N=Nc4cc(ccc4OC)C(=O)Nc5ccccc5)c2O)c(OC)cc1Cl |
| IUPAC-Name | N-(4-Chlor-2,5-dimethoxyphenyl)-3-hydroxy-4-[(E)-[2-methoxy-5-(phenylcarbamoyl)phenyl]diazenyl]naphthalin-2-carboxamid |
| Molekulargewicht (g/mol) | 611.04 |
| Formelmasse | 610.1619 |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?