Learn More
PIGN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559149
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KVDDGVKEIV SMFNHFYGND GKTTFIFTSD HGMTDWGSHG AGHPSETLTP LVTWGAGIKY PQRVSAQQFD DAFLKEWRLE NWK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
PIGN Polyclonal antibody specifically detects PIGN in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
PIGN | |
Polyclonal | |
Unconjugated | |
PIGN | |
Gm20308; GPI ethanolamine phosphate transferase 1; MCAHS; MCAHS1; MCD4; MCD4 homolog; MDC4; phosphatidylinositol glycan anchor biosynthesis class N; phosphatidylinositol glycan anchor biosynthesis, class N; phosphatidylinositol glycan, class N; phosphatidylinositol-glycan biosynthesis class N protein; PIGN; PIG-N | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95427 | |
PIGN | |
Recombinant protein corresponding to Human PIGN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only