Learn More
PIK3R6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554689
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PAAPVTDHTC LNVNVTEVVK SSNLAGKSFS TVTNTFRTNN IQIQSRDQRL LTLSLDKDDQ RTFRDVVRFE VAPCPEPCSG AQKSKAPWLN LHGQQEVEAI KAKPKP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 67%.
PIK3R6 Polyclonal antibody specifically detects PIK3R6 in Human samples. It is validated for Immunohistochemistry.Spezifikation
PIK3R6 | |
Polyclonal | |
Unconjugated | |
PIK3R6 | |
BB220380; C17orf38; HsT41028; p84; p84 PI3K adapter protein; p84 PIKAP; p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein; p87 phosphoinositide 3-kinase gamma adapter; p87 PI3K adapter protein; p87(PIKAP); p87PIKAP; Phosphoinositide 3-kinase gamma adapter protein of 87 kDa; Phosphoinositide 3-kinase regulatory subunit 6; phosphoinositide-3-kinase regulatory subunit 6; phosphoinositide-3-kinase, regulatory subunit 6; PI3Kgamma adapter protein of 87 kDa; PIK3R6; RGD1560850 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146850 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5UE93 | |
PIK3R6 | |
Recombinant protein corresponding to Human PIK3R6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only