Learn More
PITPNB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551403
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CSVQEYQVGQ LYSVAEASKN ETGGGEGIEV LKNEPYEKDG EKGQYTHKIY HLKSKVPAFV RMIAPEGSLV FHEKAWNAYP YCRTIVTNEY MKDDFFIKIE TWHKPDLGTL ENVHGLDPNT Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
PITPNB Polyclonal antibody specifically detects PITPNB in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PITPNB | |
Polyclonal | |
Unconjugated | |
PITPNB | |
phosphatidylinositol transfer protein beta; phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; Pitp beta; PI-TP beta; PI-TP-beta; Pitpnb; ptdIns transfer protein beta; PtdInsTP; ptdInsTP beta; VIB1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114561, 23760, 56305 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P48739, P53811, P53812 | |
PITPNB | |
Recombinant protein corresponding to Human PITPNB | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only