missing translation for 'onlineSavingsMsg'
Learn More

PKLR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 17580-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887504

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 25-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The protein encoded by this gene is a pyruvate kinase that catalyzes the transphosphorylation of phohsphoenolpyruvate into pyruvate and ATP, which is the rate-limiting step of glycolysis. Defects in this enzyme, due to gene mutations or genetic variations, are the common cause of chronic hereditary nonspherocytic hemolytic anemia (CNSHA or HNSHA). Multiple transcript variants encoding different isoforms have been found for this gene.
Spezifikation

Spezifikation

PKLR
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P12928, P30613, P53657
PK1, PKL, PKLR, PKR, PKRL, Pyruvate kinase 1, Pyruvate kinase isozymes R/L, pyruvate kinase, liver and RBC, R type/L type pyruvate kinase, Red cell/liver pyruvate kinase, RPK
Rabbit
IgG
150 μL
-20°C
Primary
18770, 24651, 5313
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PKLR
Liquid
Pklr
PKLR Fusion Protein Ag11096
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate