Learn More
PKM Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Artikelnummer. p-7087698 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16896223 20 μL 20 Mikroliter
16886223 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16896223 Lieferant Proteintech Lieferanten-Nr. 100782AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
This antibody can recognize both PK-M1 and PK-M2.
PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.
Spezifikation
Antigen PKM
Anwendungen Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 0.14 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gen PKM
Gen-Zugriffsnummer P14618, P52480
Gen-Alias CTHBP, OIP 3, OIP3, Opa interacting protein 3, p58, PK2, PK3, PKM, PKM1, PKM2, PKM2 pyruvate kinase, muscle, Pyruvate kinase 2/3, Pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, TCB, THBP1, Tumor M2 PK
Gensymbole PKM
Wirtsspezies Rabbit
Immunogen PKM Fusion Protein Ag0110
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 18746, 5315
Zielspezies Mouse, Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG