missing translation for 'onlineSavingsMsg'
Learn More

PKM Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10078-2-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886223

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody can recognize both PK-M1 and PK-M2.

PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.
Spezifikation

Spezifikation

PKM
0.14 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14618, P52480
CTHBP, OIP 3, OIP3, Opa interacting protein 3, p58, PK2, PK3, PKM, PKM1, PKM2, PKM2 pyruvate kinase, muscle, Pyruvate kinase 2/3, Pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, TCB, THBP1, Tumor M2 PK
Rabbit
IgG
150 μL
-20°C
Primary
18746, 5315
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PKM
Liquid
PKM
PKM Fusion Protein Ag0110
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation