missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550637
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total PLA2G6 protein.
PLA2G6 Polyclonal antibody specifically detects PLA2G6 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
PLA2G6 | |
Polyclonal | |
Unconjugated | |
PLA2G6 | |
85 kDa calcium-independent phospholipase A2; 85/88 kDa calcium-independent phospholipase A2; BB112799; CaI-PLA2; Group VI phospholipase A2; GVI; GVI PLA2; INAD1; intracellular membrane-associated calcium-independent phospholipase A2 beta; iPLA(2)beta; iPLA2; iPLA2beta; iPLA2-beta; IPLA2-VIA; NBIA2; NBIA2A; NBIA2B; neurodegeneration with brain iron accumulation 2; PARK14; patatin-like phospholipase domain-containing protein 9; phospholipase A2 group VI; phospholipase A2, group VI; phospholipase A2, group VI (cytosolic, calcium-independent); PLA2; PLA2G6; PLPLA9; PNPLA9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
360426, 53357, 8398 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O60733, P97570, P97819 | |
PLA2G6 | |
Synthetic peptide corresponding to a region derived from internal residues of human phospholipase A2, group VI (cytosolic, calcium-independent) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |