Learn More
PLAA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554300
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EGKEFDYVFS IDVNEGGPSY KLPYNTSDDP WLTAYNFLQK NDLNPMFLDQ VAKFIIDNTK GQMLGLGNPS FSDPFTGGGR YVPGSSGSSN TLPTADP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
PLAA Polyclonal antibody specifically detects PLAA in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLAA | |
Polyclonal | |
Unconjugated | |
Plaa | |
2410007N06; AI536418; AU018445; AW208417; D4Ertd618e; DOA1; DOA1 homolog; phospholipase A2 activating protein; phospholipase A2, activating protein; phospholipase A-2-activating protein; phospholipase A2-activating protein; PLA2P; Plaa; PLAP; Ufd3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116645, 18786, 9373 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P27612, P54319, Q9Y263 | |
Plaa | |
Recombinant protein corresponding to Human PLAA | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only