missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCB4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567835
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with Mouse and Rat samples.
The production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes. This form has a role in retina signal transduction.Spezifikation
PLCB4 | |
Polyclonal | |
Unconjugated | |
PLCB4 | |
1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-4; A930039J07Rik; AI854601; ARCND2; Beta4aa; C230058B11Rik; dJ1119D9.2 (Phopholipase C, beta 4 (1-Phosphatidylinositol-4,5-Bisphosphate Phosphodiesterase Beta 4)); inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; PCL-C1; phosphoinositidase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-beta-4; phosphoinositide-specific phospholipase c; phospholipase C; phospholipase C beta 4; phospholipase C, beta 4; phospholipase C-beta-4; PI-PLC; PLCB4; PLC-beta-4; RATBETA4AA; triphosphoinositide phosphodiesterase | |
Rabbit | |
affinity chromatography | |
RUO | |
18798, 25031, 5332 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15147, Q9QW07 | |
PLCB4 | |
Synthesized peptide derived from human PLCB4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |