Learn More
PLD6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561939
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QDPGYMHHKF AIVDKRVLIT GSLNWTTQAI QNNRENVLIT EDDEYVRLFL EEFERIWEQF NPTKYTFFPP KKSHGSCAPP VSRAGGRLLS WHRTCGTSSE SQT Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 61%.
PLD6 Polyclonal antibody specifically detects PLD6 in Human samples. It is validated for Immunohistochemistry.Spezifikation
PLD6 | |
Polyclonal | |
Unconjugated | |
PLD6 | |
4933433K01Rik; choline phosphatase 6; Gm10; Mitochondrial cardiolipin hydrolase; mitochondrial phospholipase; mitoPLD; mZuc; novel Phospholipase D Active site motif-containing protein; phosphatidylcholine-hydrolyzing phospholipase D6; phospholipase D family member 6; phospholipase D family, member 6; phospholipase D6; PLD 6; PLD6; Protein zucchini homolog; Zuc | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201164 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N2A8 | |
PLD6 | |
Recombinant protein corresponding to Human PLD6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only