Learn More
PLEKHA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561748
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RTQAHSGSPT YLQLPPRPPG TRASMVLLPG PPLESTFHQS LETDTLLTKL CGQDRLLRRL QEEIDQKQEE KEQLEAALEL TRQQLGQA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
Binds specifically to phosphatidylinositol 3-phosphate (PtdIns3P), but not to other phosphoinositides.Spezifikation
PLEKHA4 | |
Polyclonal | |
Unconjugated | |
PLEKHA4 | |
PEPP1; PEPP-1; PH domain-containing family A member 4; phosphoinositol 3-phosphate binding protein-1; phosphoinositol 3-phosphate-binding PH domain protein 1; phosphoinositol 3-phosphate-binding protein 1; pleckstrin homology domain containing A4; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 4; pleckstrin homology domain-containing family A member 4; PLEKHA4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57664 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4M7 | |
PLEKHA4 | |
Recombinant protein corresponding to Human PLEKHA4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only