Learn More
PLEKHA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558465
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SSLKRDMEKV ERQAVPQANH TESCHECGRV GPGHTRDCPH RGHDDIVNFE RQEQEGEQYR SQRDPLEGKR DRSKARSPYS PAEEDALFMD LPTGP Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.
PLEKHA7 Polyclonal antibody specifically detects PLEKHA7 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLEKHA7 | |
Polyclonal | |
Unconjugated | |
PLEKHA7 | |
A430081P20Rik; AI414265; DKFZp686M22243; Hadp1; Heart adapter protein 1; PH domain-containing family A member 7; PH domain-containing family A member 7 antibody; pleckstrin homology domain containing A7; pleckstrin homology domain containing A7 antibody; pleckstrin homology domain containing, family A member 2; pleckstrin homology domain containing, family A member 7; pleckstrin homology domain-containing family A member 7; Plekha7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IQ23 | |
PLEKHA7 | |
Recombinant protein corresponding to Human PLEKHA7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only