missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15410-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Cargo transport protein that is required for apical transport from the Golgi complex. Transports AQP2 from the trans-Golgi network (TGN) to sites of AQP2 phosphorylation. Mediates the non-vesicular transport of glucosylceramide (GlcCer) from the trans-Golgi network (TGN) to the plasma membrane and plays a pivotal role in the synthesis of complex glycosphingolipids. Binding of both phosphatidylinositol 4-phosphate (PIP) and ARF1 are essential for the GlcCer transfer ability. Also required for primary cilium formation, possibly by being involved in the transport of raft lipids to the apical membrane, and for membrane tubulation.Spezifikation
PLEKHA8 | |
Polyclonal | |
Unconjugated | |
PLEKHA8 | |
FAPP 2, FAPP2, hFAPP2, PLEKHA8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231999, 500132, 84725 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZY60, Q80W71, Q96JA3 | |
Plekha8 | |
PLEKHA8 Fusion Protein Ag7677 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |