Beschreibung
Guanine nucleotide exchange factor that activates RHOA and maybe the NF-kappa-B signaling pathway. Involved in the control of neuronal cell differentiation. Plays a role in angiogenesis through regulation of endothelial cells chemotaxis.
Spezifikation
Spezifikation
| Antigen | PLEKHG5 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.29 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | PLEKHG5 |
| Gen-Zugriffsnummer | O94827, Q66T02, Q6RFZ7 |
| Gen-Alias | DSMA4, GEF720, KIAA0720, PLEKHG5, RP4 650H14.3 |
| Gensymbole | Plekhg5 |
| Mehr anzeigen |