Learn More
PLEKHG6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559578
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GLLMEVSAET LFGNVPSLIR THRSFWDEVL GPTLEETRAS GQPLDPIGLQ SGFLTFGQRF HPYVQYCLRV KQTMAYAREQ QETNPLFHAF VQWCE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.
PLEKHG6 Polyclonal antibody specifically detects PLEKHG6 in Human samples. It is validated for Immunohistochemistry.Spezifikation
PLEKHG6 | |
Polyclonal | |
Unconjugated | |
PLEKHG6 | |
BC026778; Gm155; MyoGEF; myosin II interacting GEF; myosin interacting guanine nucleotide exchange factor; myosin-interacting guanine nucleotide exchange factor; PH domain-containing family G member 6; pleckstrin homology and RhoGEF domain containing G6; pleckstrin homology domain containing, family G (with RhoGef domain) member 6; pleckstrin homology domain-containing family G member 6; PLEKHG6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55200 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3KR16 | |
PLEKHG6 | |
Recombinant protein corresponding to Human PLEKHG6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only