missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLK3, GST Tag Recombinant Protein
PLK3 is a member of the polo family of serine/threonine kinases.
Marke: Invitrogen™ PR7316B
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PLK3 is involved in the regulation of M phase in the cell cycle.
Spezifikation
NP_004064 | |
59.7 kDa | |
PLK3, CNK | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: Other | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
100μg | |
PLK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |