missing translation for 'onlineSavingsMsg'
Learn More

PLUNC Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10413-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16886783

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 04-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PLUNC is the human homolog of murine plunc and is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this protein is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. This gene is the human homolog of murine plunc, and like the mouse gene, is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this gene is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. Multiple transcript variants resulting from alternative splicing in the 3′ UTR have been detected, but the full-length nature of only two is known.
Spezifikation

Spezifikation

PLUNC
Polyclonal
Unconjugated
BPIFA1
LPLUNC3, Lung specific protein X, LUNX, NASG, PLUNC, Protein Plunc, SPLUNC1, SPURT, Von Ebner protein Hl
Rabbit
Antigen Affinity Chromatography
RUO
18843, 51297
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P97361, Q9NP55
BPIFA1
PLUNC Fusion Protein Ag0629
20 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote