Learn More
PMPCB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559246
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: WGFSESLLIR GAAGRSLYFG ENRLRSTQAA TQVVLNVPET RVTCLESGLR VASEDSGLST CTVGLWIDAG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
PMPCB Polyclonal antibody specifically detects PMPCB in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
PMPCB | |
Polyclonal | |
Unconjugated | |
PMPCB | |
3110004O18Rik; Beta-MPP; mitochondrial processing peptidase beta subunit; mitochondrial-processing peptidase subunit beta; MPP11; MPPB; MPPP52; P-52; peptidase (mitochondrial processing) beta; peptidase, mitochondrial processing beta subunit; PMPCB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75439 | |
PMPCB | |
Recombinant protein corresponding to Human PMPCB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only