Learn More
Macherey-Nagel™ POLYGRAM-Fertigfolien CEL 300 Format: 4 x 8 cm (VE=4x50Stck.)
Alle ansehen Macherey-Nagel ProdukteBeschreibung
TLC polyester sheets POLYGRAM CEL 300, native fibrous cellulose layer.
Spezifikation
Spezifikation
| Material | Polyesterplatten Polygram CEL 300 |
| Menge | 200er-Packung |
| Typ | TLC Micro-Set M |
| Enthält | 2 developing chambers; 2 x 50 glass capillaries 1 μL; 1 rubber cap for glass capillaries; 2 spotting guides; 1 measuring cylinder 10 mL; 1 beaker 25 mL; 1 glass laboratory sprayer with rubber bulb; 1 plastic syringe 1 mL; 20 Sheets filter paper MN 713, 15 x 21 cm; 50 polyester sheets Polygram CEL 300 |
| Abmessungen (L x B) | 4 x 8 cm |