Learn More
Polyethylene Glycol 3350, Powder, BAKER™, J.T. Baker™
Catalog Number p-5391322
Click to view available options
Menge:
1 kg
2 kg
Verpackung:
Poly Pail
Wide Mouth Amber Glass Bottle
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO
A grade of chemicals for laboratory use
Chemischer Name oder Material Polyethylene Glycol 3350
Physikalische Form Powder
Menge 1 kg
Formelmasse 3000 to 3700 (Average)
Verpackung Wide Mouth Amber Glass Bottle