missing translation for 'onlineSavingsMsg'
Learn More

Polyethylenglycol Dimethacrylat, Molekulargewicht 8,000, Thermo Scientific Chemicals

Catalog Number 15467355
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
25852-47-5
MFCD00081877
ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
25852-47-5
MFCD00081877
ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate

Polyethylene glycol dimethacrylate is used as adhesives, coatings, sealants, photo resists, solder masks and photopolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Polyethylene glycol dimethacrylate
Farbe Weiß
Namenshinweis M.W. 8,000
Physikalische Form Granularer Feststoff
Summenformel CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Löslichkeitsinformationen Soluble in water.
Menge 250 mg
Empfindlichkeit Light and moisture sensitive

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'