missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Waschflaschen aus PP (Polypropylen)
Click to view available options
Kapazität (metrisch):
1000 ml
Beschreibung
- Bottle, screw cap, and spray tube made from polypropylene
- A pressure relief valve in the screw cap stops discharge, dripping or leakage of liquid through the spray tube due to temperature fluctuations
- Chemical resistance of the bottles are limited when aggressive filling media are used
- Rated for Acetone
Spezifikation
Spezifikation
Kapazität (metrisch) | 1000 ml |
Enthält | Spray tube, pressure relief valve |
Material | Polypropylen |
Anzahl pro Packung | Je 1 |
Closure Included | Ja |
Produkttyp | Spülflasche |
Chemical Label | Aceton |
Verschlussmaterial | Polypropylen |
Verschlusstyp | Schraubverschluss |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'