Learn More
Carl Roth™ Polystyrene Flexistrip Spreading Spatula
Marke: Carl Roth™ L430.1
Weitere Details : Netto-Gewicht : 1.00000kg
Spezifikation
1 Pk. | |
Uniform spreading of blood smears on lens carriers | |
75 x 26 mm | |
75 mm |
Flexistrip Spreading Spatula | |
26 mm | |
Polystyrene |