missing translation for 'onlineSavingsMsg'
Learn More

Kalium-1-Naphthalintrifluorborat, 96 %, Thermo Scientific™

Catalog Number p-7035265
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
166328-07-0
C10H7BF3K
234.07
MFCD06659936
SBEPDIKXYGSGJL-UHFFFAOYSA-N
potassium 1-naphthalene trifluoroborate, potassium trifluoro naphthalen-1-yl borate, potassium trifluoro naphthalen-1-yl boranuide, potassium 1-naphthalenetrifluoroborate, amtb547, 1-naphthylpotassiotrifluoroboron v, potassium trifluoro naphthalen-1-yl borate 1-, borate 1-,trifluoro-1-naphthalenyl-,potassium 1:1 , t-4
23670988
Kalium;Trifluoro(Naphthalen-1-yl)Boranuid
[B-](C1=CC=CC2=CC=CC=C12)(F)(F)F.[K+]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
166328-07-0
C10H7BF3K
234.07
MFCD06659936
SBEPDIKXYGSGJL-UHFFFAOYSA-N
potassium 1-naphthalene trifluoroborate, potassium trifluoro naphthalen-1-yl borate, potassium trifluoro naphthalen-1-yl boranuide, potassium 1-naphthalenetrifluoroborate, amtb547, 1-naphthylpotassiotrifluoroboron v, potassium trifluoro naphthalen-1-yl borate 1-, borate 1-,trifluoro-1-naphthalenyl-,potassium 1:1 , t-4
23670988
Kalium;Trifluoro(Naphthalen-1-yl)Boranuid
[B-](C1=CC=CC2=CC=CC=C12)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Potassium 1-naphthalenetrifluoroborate
Schmelzpunkt 89°C to 91°C
Menge 1 g
Formelmasse 234.07
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'