missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kaliumbromat, 99.8 % min., Thermo Scientific Chemicals
Click to view available options
Menge:
100 g
500 g
Beschreibung
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Chemischer Name oder Material | Potassium bromate |
Schmelzpunkt | 370°C (decomposition) |
Physikalische Form | granular |
Menge | 100 g |
Prozentgehaltsbereich | 99.8% min |
Löslichkeitsinformationen | Slightly soluble in alcohol but insoluble in acetone and ethanol. |
Formelmasse | 167 |
Güte | ACS-Reagenz |
Dichte | 3.27 g/mL |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'