missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Potassium hydride, 30% w/w in mineral oil

Product Code. 18982371
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
18982371 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18982371 Supplier Thermo Scientific™ Supplier No. L13266.NB
 Restricted Item
Lager vorrätig
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7693-26-7 | HK | 40.11 g/mol

Potassium hydride is a precursor in the production of various bases by deprotonation, highly-hindered bases potassium triethylmethoxide and potassium 3-aminopropylamide It serves as a powerful base in organic synthesis of deprotonate carbonyl compounds and amines to form enolates and amides respectively.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7693-26-7
Density 1.45
Quantity 5 g
Flash Point 160°(320°F)
Form Liquid
Molecular Formula HK
MDL Number MFCD00011357
UN Number UN1409
Fieser 1,935 12,407 13,257 14,265 17,290 18,299 19,276 21,360
InChI Key OCFVSFVLVRNXFJ-UHFFFAOYSA-N
SMILES [H-].[K+]
IUPAC Name potassium hydride
Molecular Weight (g/mol) 40.11
Formula Weight 40.11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.