missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Semiconductor grade, Honeywell
Click to view available options
Menge:
100 g
500 g
Spezifikation
Chemischer Name oder Material | Potassium Hydroxide |
Strukturformel | KOH |
UN-Nummer | 1813 |
Menge | 100 g |
Formelmasse | 56.11g/mol |
Reinheit (%) | 99.99% (trace metals basis) |
Güte | Semiconductor |
Dampfdruck | 1mmHg (719°C) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'