Learn More
IBA Lifesciences™ PPI (2-10) Peptide ALWMRLLPL (HLA-A*0201)
Alle ansehen IBA Lifesciences ProdukteBeschreibung
PPI (2-10) peptide ALWMRLLPL (HLA-A*0201) is a single peptide for stimulation of T cells. The peptide from preproinsulin (PPI) is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- Peptides have been purified by HPLC using a water and acetonitrile gradient
- Endotoxin-free
Spezifikation
Spezifikation
| Reinheit | >85% |
| Endotoxin-Gehalt | Endotoxin-free |
| Zur Verwendung mit (Anwendung) | T Cell Stimulation |
| Lyophilisiert | Yes |
| Menge | 1 mg |