missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3F Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55386-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Glycogen-targeting subunit for protein phosphatase 1 (PP1).Spezifikation
PPP1R3F | |
Polyclonal | |
Unconjugated | |
PPP1R3F | |
Hb2E, PPP1R3F | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54646, 89801 | |
-20°C | |
Liquid |
Western Blot | |
0.76 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZSY5, Q9JIG4 | |
Ppp1r3f | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |