Learn More
Advantec Mfs™ Druckbehälter
Pressure vessels allow for the dispensing of process liquids through a filter holder via pressure filtration. Available in 304 or 316 stainless steel, electropolished for a smooth, cleanable surface. Choose a standard mouth or wide mouth opening for ease of cleaning.
N.V.
Spezifikation
Zertifizierung/Konformität | ASME Certified; certificate of analysis available on request |
---|---|
Enthält | Hauptgefäß und -boden, Verschlusseinheit und Tauchschlauch-Einheit |
Öffnung | Wide mouth opening |
Menge | Gehäuse 1Pack |
Beschreibung
- Available in 304 or 316 stainless steel
- Electropolished finish provides cleanable surface
- Standard opening or wide mouth opening for ease of cleaning
- Removable molded rubber foot protects vessel base
- 316 stainless steel complies with current GMP and is recommended for low pH and high salt applications
- Optional vacuum closure is recommended when working with negative pressure or when operating at low pressure (<30 psi) or with stiffer O-ring material (PTFE)
- ASME Certified
Spezifikation
ASME Certified; certificate of analysis available on request | |
Hauptgefäß und -boden, Verschlusseinheit und Tauchschlauch-Einheit | |
Wide mouth opening | |
Gehäuse 1Pack |