Learn More
Advantec Mfs™ Druckbehälter
Pressure vessels allow for the dispensing of process liquids through a filter holder via pressure filtration. Available in 304 or 316 stainless steel, electropolished for a smooth, cleanable surface. Choose a standard mouth or wide mouth opening for ease of cleaning.
N.V.
Spezifikation
Zertifizierung/Konformität ASME Certified; certificate of analysis available on request
Enthält Hauptgefäß und -boden, Verschlusseinheit und Tauchschlauch-Einheit
Öffnung Wide mouth opening
Menge Gehäuse 1Pack
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
  • Available in 304 or 316 stainless steel
  • Electropolished finish provides cleanable surface
  • Standard opening or wide mouth opening for ease of cleaning
  • Removable molded rubber foot protects vessel base
  • 316 stainless steel complies with current GMP and is recommended for low pH and high salt applications
  • Optional vacuum closure is recommended when working with negative pressure or when operating at low pressure (<30 psi) or with stiffer O-ring material (PTFE)
  • ASME Certified
Spezifikation
Spezifikation
ASME Certified; certificate of analysis available on request
Hauptgefäß und -boden, Verschlusseinheit und Tauchschlauch-Einheit
Wide mouth opening
Gehäuse 1Pack