Learn More
Tocris Bioscience™ Pristimerin
Potent, reversible MAGL inhibitor
N.V.
Beschreibung
Potent and reversible inhibitor of monoacylglycerol lipase (MGL) (IC50 = 93 nM). Also suppresses NF-κB activation via inhibition of proteasome chymotrypsin-like activity and IKKαβ. Displays antitumor, anti-inflammatory and antimicrobial activities.Spezifikation
>99% |
For Research Use Only