missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17628-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.Spezifikation
PRMT3 | |
Polyclonal | |
Unconjugated | |
PRMT3 | |
HRMT1L3, PRMT3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10196, 71974, 89820 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60678, O70467, Q922H1 | |
Prmt3 | |
PRMT3 Fusion Protein Ag11877 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |