missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Prohibitin Monoclonal antibody specifically detects Prohibitin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Spezifikation
Spezifikation
| Antigen | Prohibitin |
| Anwendungen | Western Blot, Immunohistochemistry (Paraffin), Protein Array |
| Klassifikation | Monoclonal |
| Klon | PHB/3228 |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | PHB1, prohibitin |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary) (Uniprot: P35232) |
| Reinigungsverfahren | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?