Learn More
Propyne, ca. 4% in N,N-Dimethylformamide, ca. 1.0mol/L, TRC
Product Code. 30427957
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427957 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30427957 Supplier Toronto Research Chemicals Supplier No. TRCP83598525ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Propyne
CAS 74-99-7
Molecular Formula C3H4
Quantity 25 mL
Synonym Allylene, Methylacetylene, Propine
InChI Formula InChI=1S/C3H4/c1-3-2/h1H,2H3
SMILES CC#C
IUPAC Name prop-1-yne
Molecular Weight (g/mol) 40.06
Formula Weight 40.0313
Concentration or Composition (by Analyte or Components) 4 %