missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Proteinase K Recombinant Protein
Yeast Recombinant Protein
Marke: Invitrogen™ RP87705
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Proteinase K (E.C.3.4.21.64) is an extracellular serine endoproteinase produced by Tritirachinm album Limber. Proteinase K is active in a wide range of temperatures and buffers with optimal activity between 20 and 6°C and a pH between 7.5 and 12.0. Activity is stimulated when up to 2% SDS or up to 4 M urea are included in the reaction. Calcium is important for thermostability of Proteinase K but it is not required for catalysis, therefore Proteinase K is also active in buffers containing chelating agents such as EDTA.- Purity: > 95% by SDS-Page
- Protein Length/ AA Sequence: full length Proteinase K protein
- Biological Activity: (Measured by its digestion activity. The activity is >30U/mg. One unit will digest Casein at 37°C (pH 7.5) per minute to produce equal absorbance as 1.0 μmol of L-tyrosine.
- Animal Origin Free: Yes No DNase contamination detected by digestion of lambda DNA at 37°C for 6 hours. No RNases contamination detected by digestion of lambda DNA at 25°C for 16 hours. Host genomic DNA/RNA is no residual detected by PCR. Storage: Store the lyophilized protein at 4°C for up to 1 year and less than -20°C for long term. Lyophilized powder is recommended to dissolve in in 20mM Tris-Hcl, 30mM NaCl, 50% Glycerol, 5mM CaCl2, pH 7.2-7.4. After reconstitution, the protein solution is stable at -20°C for 1 year, at 2-8°C for up to 6 months.
Spezifikation
Control, ELISA, Western Blot | |
Proteinase K | |
Yeast | |
RUO | |
Fungi | |
Recombinant | |
>95% |
protein with no preservative; pH 7.4 | |
1 g | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
EC 3.4.21.14; Protease K | |
Unconjugated | |
Lyophilized |