missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocadherin FAT2 Mouse, Clone: 8C5, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 2196-MSM1-P1
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Protocadherin FAT2 | |
Cell Surface | |
8C5 | |
Unconjugated | |
XP_016864713 | |
Mouse | |
480kDa | |
Protein A/G Purified | |
RUO | |
2196 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
FAT2 | |
Purified recombinant human FAT2 fusion protein | |
Esophagus, Cervix, Uterus, Tonsil or Cerebellum. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |