missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRPS2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 13299088
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
13299088 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 13299088 Lieferant Invitrogen™ Lieferanten-Nr. PA521537

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (97%), Xenopus laevis (96%), Pig (97%), Chicken (96%), Chimpanzee (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Phosphoribosyl Pyrophosphate Synthetase 2 (PRPS2) gene plays a critical role in cellular metabolism by encoding an enzyme that conducts a key reaction linking central carbon metabolism and nucleotide production, essential for various biological functions. Structurally, PRPS2 shares a high sequence identity with PRPS1, making it less sensitive to allosteric inhibition and particularly significant in certain oncology contexts due to its upregulation at the translational level in some cancers. The enzyme encoded by PRPS2 is pivotal in coupling protein and nucleotide biosynthesis, aided by a unique cis-regulatory element in its 5' UTR, which is influenced by oncogenic pathways. This gene manifests a tissue-specific expression profile, predominantly in proliferative tissues such as the thymus, lung, and small intestine, highlighting its significance in cell growth and proliferation. The evolutionary origins of PRPS2, along with its functional diversification into multiple PRPS genes in eukaryotes, underscore its indispensable role in cellular physiology and pathology.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen PRPS2
Anwendungen Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.91 mg/mL
Konjugat Unconjugated
Zusammensetzung 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gen PRPS2
Gen-Zugriffsnummer P11908
Gen-Alias 2610101M19Rik; AA589463; AI464149; Phophoribosylpyrophosphate synthetase, subunit II; phosphoribosyl pyrophosphate synthase II; phosphoribosyl pyrophosphate synthetase 2; phosphoribosyl pyrophosphate synthetase II; PPRibP; PPRibP synthetase; Prps2; Prps-2; PRSII; PRS-II; ribose-phosphate diphosphokinase 2; ribose-phosphate pyrophosphokinase 2; testicular secretory protein Li 41
Gensymbole PRPS2
Wirtsspezies Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRPS2. The exact sequence is proprietary.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 5634
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.