missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSMC4 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15749630 Alle ansehen Thermo Scientific Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15749630 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15749630 Lieferant Invitrogen™ Lieferanten-Nr. PA551676

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: LEDLYSRYKK LQQELEFLEV QEEYIKDEQK NLKKEFLHAQ EEVKRIQSIP LVIGQFLEAV DQNTAIVGST TGSNYYVRIL STIDRELLKP NASVALHKHS NA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the ATPase subunits, a member of the triple-A family of ATPases which have a chaperone-like activity. This subunit has been shown to interact with an orphan member of the nuclear hormone receptor superfamily highly expressed in liver, and with gankyrin, a liver oncoprotein. Two transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen PSMC4
Anwendungen Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen PSMC4
Gen-Zugriffsnummer P43686, P54775, Q63570
Gen-Alias 26S protease regulatory subunit 6B; 26S proteasome AAA-ATPase subunit RPT3; 26S proteasome regulatory subunit 6B; CAR interacing protein 21; CAR interacting protein 21; CIP21; MB67-interacting protein; MGC13687; MGC23214; MGC8570; MIP224; protease 26S subunit 6; proteasome (prosome, macropain) 26S subunit, ATPase, 4; proteasome 26S ATPase subunit 4; proteasome 26S subunit ATPase 4; proteasome 26S subunit, ATPase 4; PSMC4; RPT3; S6; S6 ATPase; Tat-binding protein 7; TBP7; TBP-7
Gensymbole PSMC4
Wirtsspezies Rabbit
Immunogen Recombinant protein corresponding to Human PSMC4. Recombinant protein control fragment (Product #RP-102166).
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 117262, 23996, 5704
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.