missing translation for 'onlineSavingsMsg'
Learn More

PSMC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 14752-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16883554

  • 112.71 € / 20 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.
Spezifikation

Spezifikation

PSMC5
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P62195, P62196, P62198
p45, p45/SUG, Proteasome subunit p45, PSMC5, Rpt6, S8, SUG1, TBP10, TRIP1
Rabbit
IgG
20 μL
-20°C
Primary
19184, 5705, 81827
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PSMC5
Liquid
Psmc5
PSMC5 Fusion Protein Ag6497
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation