missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSRC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 27130-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a proline-rich protein. Studies of this gene and the related mouse gene suggest that this gene is regulated by p53 and may participate in p53-mediated growth suppression. The encoded protein may function as a microtubule destabilizing protein that controls spindle dynamics and mitotic progression by recruiting and regulating microtubule depolymerases. At least one genetic variation in this gene has been associated with decreased serum levels of low-density lipoprotein cholesterol. Alternatively spliced transcript variants encoding different isoforms have been described.Spezifikation
PSRC1 | |
Polyclonal | |
Unconjugated | |
PSRC1 | |
DDA3, FP3214, PSRC1, RP11 297O4.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84722 | |
-20°C | |
Liquid |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PGN9 | |
PSRC1 | |
PSRC1 Fusion Protein Ag26039 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |