Learn More
Cole-Parmer™ PROPELLERRUEHRER PTFE 460X8
Marke: Cole-Parmer™ 04449-10
Weitere Details : Netto-Gewicht : 0.80000kg
Spezifikation
50.8 mm | |
Use these Plastic paddle assemblies for mixing most acids and corrosive solutions | |
1 Each |
7.9375 mm | |
457.2 mm | |
Propeller |