Learn More
Pyrex™ Vakuum-Filternutsche aus Borosilikatglas mit Frittenboden aus Glas
Useful when conventional filter papers react with chemicals.
Marke: Quickfit VB40/06M
Weitere Details : Netto-Gewicht : 0.19000kg
Beschreibung
- Büchner vacuum filtration funnel with glass sintered disc
- Manufactured from borosilicate glass for highest resistance to chemical attack
- Useful where conventional filter papers would be destroyed by acids and alkalis
- With a Quickfit™ ground glass cone to allow direct filtration into Quickfit™ Büchner flasks